Cole-Parmer® Stuart WS-200 Water Stills
Lieferant: Cole Parmer
99295-00 99295-01
STUA99295-00EA 4730 CHF
STUA99295-00 STUA99295-01
Cole-Parmer® Stuart WS-200 Water Stills
Wasseraufbereitungssysteme Wasserdestillierapparate
Get high purity water with all-glass construction.
  • Two independent safety thermostats prevent overheating
  • Heating element shuts down when reservoir is full
This all-glass water still with silica-sheathed heaters and high-efficiency condenser ensures top distillate purity at an economical price. A reservoir level control feature automatically turns off the heater when the reservoir is full preventing any overflow. Two independent safety thermostats, mounted in the condenser and boiler, prevent overheating in the event the water supply is interrupted.
More Product Information