Polyethylenglycol 300

Lieferant: Spectrum Chemical

Synonyms: PEG 300

PO108-2.5LTGL PO108-200LTBL PO108-20LT PO108-500MLGL
SPCMPO108-2.5LTGLEA 500 CHF
SPCMPO108-2.5LTGL SPCMPO108-200LTBL SPCMPO108-20LT SPCMPO108-500MLGL
Polyethylenglycol 300
Polyethylenglycol 300

Polyethylene Glycol 300, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formel: H(OCH₂CH₂)nOH
MW: 300 g/mol
Flash-Pt: 220
MDL Number: MFCD00081839
CAS-Nummer: 25322-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR