Phenol 99,0, crystals_fused USP

Lieferant: Spectrum Chemical

Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene

PH120-1.5KGPL PH120-25KGBL PH120-500GMGL
SPCMPH120-1.5KGPLEA 981 CHF
SPCMPH120-1.5KGPL SPCMPH120-25KGBL SPCMPH120-500GMGL
Phenol 99,0, crystals_fused USP
Phenol

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: C₆H₅OH
MW: 94.11 g/mol
Siedepunkt: 181.7
Melting Pt: 40.8
Dichte: 1.071
Flash-Pt: 81
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00002143
CAS-Nummer: 108-95-2
UN: 1671
ADR: 6.1 II
Merck Index: 12,07390

Order Now

Spezifikation Testergebnisse

Assay (Anhydrous Basis) 99.0 - 100.5 %
Clarity of Solution and Reaction To Pass Test
Congealing Temperature 39°C min.
Water Determination max. 0.5 %
Nonvolatile Residue max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR