2-Phenylethanol USP

Lieferant: Spectrum Chemical
Gefahr

Synonyms: Phenethyl alcohol, Benzylcarbinol, β-Hydroxyethylbenzene, Benzyl carbinol, Benzenethanol

P1058-100GM P1058-1KG P1058-15.9KGBL
SPCMP1058-100GMEA 306 CHF
SPCMP1058-100GM SPCMP1058-1KG SPCMP1058-15.9KGBL
2-Phenylethanol USP
2-Phenylethanol

BSE/TSE statement (available upon request)
Allergen statement (available upon request)

Formel: C₆H₅CH₂CH₂OH
MW: 122.17 g/mol
Siedepunkt: 432
Melting Pt: 103…105 °C
Dichte: 1.02 g/cm³ (20 °C)
Flash-Pt: 102
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00002886
CAS-Nummer: 60-12-8
EINECS: 200-456-2
UN: 2810
ADR: 6.1 III
Merck Index: 12,07370

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR