Lecithin, körnig, für die Nahrungsmittelindustrie

Lieferant: Spectrum Chemical

L1617-500GM L1617-2.5KG L1617-12KG L1617-50KGBL
SPCML1617-500GMEA 213 CHF
SPCML1617-500GM SPCML1617-2.5KG SPCML1617-12KG SPCML1617-50KGBL
Lecithin, körnig, für die Nahrungsmittelindustrie
Lecithin

Lecithin, Granular, Food Grade is used for a host of applications such as food additive, dietary supplement, stabilizing agent and dispersing agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Lager Temperatur: Raumtemperatur
MDL Number: MFCD00147406
CAS-Nummer: 8002-43-5
Merck Index: 12,05452

Order Now

Spezifikation Testergebnisse

Acetone-Insoluble Matter 97.0 %
Acid Value max.35
Hexane-Insoluble Matter max. 0.05 %
Water max. 1.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR