4,4'-Dimethoxytrityl chloride 98

Lieferant: Spectrum Chemical

Synonyms: DMT-Cl, 4,4'-Dimethoxytriphenylmethyl chloride

D2181-5KGBL
SPCMD2181-5KGBLEA 13720 CHF
SPCMD2181-5KGBL
4,4'-Dimethoxytrityl chloride 98
4,4'-Dimethoxytritylchlorid

Bitte beachten Storage temperature - HYGROSCOPIC: Keep tightly closed.

Formel: C₂₁H₁₉ClO₂
MW: 338.8 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008409
CAS-Nummer: 40615-36-9

Order Now

Spezifikation Testergebnisse

Assay Approx. 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR