1,3-Diaminopropane

Lieferant: Spectrum Chemical

Synonyms: 1,3-Propanediamine, Trimethylenediamine

D2049-200LTBL
SPCMD2049-200LTBLEA 0 CHF
SPCMD2049-200LTBL
1,3-Diaminopropane
1,3-Diaminopropan

1,3-Diaminopropane, also known as Trimethylenediamine, is isomeric with 1,2-diaminopropane and used in the synthesis of heterocycles. It is a simple diamine considered soluble in polar organic solvents.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: C₃H₁₀N₂
MW: 74.13 g/mol
Siedepunkt: 140
Melting Pt: -12
Dichte: 0,888
Flash-Pt: 48
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008228
CAS-Nummer: 109-76-2
UN: 2922
ADR: 86.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR