Calcium-D-saccharate tetrahydrate 98,5 USP

Lieferant: Spectrum Chemical

Synonyms: Calcium D-glucarate tetrahydrate

C1830-25KGBL
SPCMC1830-25KGBLEA 94380 CHF
SPCMC1830-25KGBL
Calcium-D-saccharate tetrahydrate 98,5 USP
Calcium-D-saccharate tetrahydrate

Calcium Saccharate, Tetrahydrate, USP is used as an antacid, as an antidote in carbolic acid poisoning, and as a stabiliser for calcium gluconate solution for parenteral administration. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formel: C₆H₈CaO₈.4H₂O
MW: 320.26 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00150704
CAS-Nummer: 5793-89-5

Order Now

Spezifikation Testergebnisse

Assay 98.5 - 102.0%
Specific Rotation [A]D +18.5 to +22.5°
Chloride (Cl) max. 0.07%
Sulfate max. 0.12%
Heavy Metals max. 0.002%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR