L-Aspartic acid monosodium salt monohydrate ≥98,0%

Lieferant: Spectrum Chemical

Synonyms: Sodium (S)-3-amino-3-carboxypropanoate hydrate, Sodium L-aspartate monohydrate, L-Aspartic acid sodium salt monohydrate

A1375-50KGBL
SPCMA1375-50KGBLEA 0 CHF
SPCMA1375-50KGBL
L-Aspartic acid monosodium salt monohydrate ≥98,0%
L-Aspartic acid monosodium salt monohydrate

L-Aspartic Acid Sodium Salt, Monohydrate is used as a principal neurotransmitter for fast synaptic excitation.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: C₄H₆NNaO₄·H₂O
MW: 173,11 g/mol (155,10 anhy)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00152960
CAS-Nummer: 323194-76-9

Order Now

Spezifikation Testergebnisse

Assay 98.0 - 101.0%
Specific Rotation [A]D +19.0 to +21.0
Transmittance 95.0 %
Chloride (Cl) max. 0.04 %
Ammonium (NH₄) max. 0.04 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR