Qualitix® Sterilisation Pouches
Lieferant: Socorex Isba
3210.0816 3210.1326 3210.1936 3210.3040 3210.0926 3210.2537 3210.3049
SOCO3210.0816EA 16.6 CHF
SOCO3210.0816 SOCO3210.1326 SOCO3210.1936 SOCO3210.3040 SOCO3210.0926 SOCO3210.2537 SOCO3210.3049
Qualitix® Sterilisation Pouches
Beutel Sterilisationsbeutel
Fitting any small to mid-size laboratory, medical, dental or veterinary device, the Qualitix® single use pouches provide easy and effective wrapping of items requiring sterilization.
  • Robust durable material
  • Quick content identification
  • Large self-sealing strip, easy to close
  • Chevron top seal for easy opening
  • Suitable for steam and EO gas sterilization
  • Two sterilization indicators
  • Five different sizes
Made with robust medical grade paper, the pouches tightly close with self-sealing strip to maintain sterility. Content remains always visible through PET/More Product Information