Water Disinfector, WD4060R
Lieferant: SMEG
WD4060R
SMEGWD4060REA 12740 CHF
SMEGWD4060R
Water Disinfector, WD4060R
Wasseraufbereitungssysteme Reinstwassersysteme
WD4060 is a washer disinfector designed to work with two to four washing levels (up to 8 cskdin basket). The washing chamber measures 530×510×620 mm (150 litres) and has the capability of washing, chemical disinfection, thermal disinfection (up to 95 °C) and active drying with hepa filter.