Sterilin® Dippas® Milk Dippers
Lieferant: STERILIN
191 192 191BLUE 194IWBLUE 192BLUE
SLIN191EA 439 CHF
SLIN191 SLIN192 SLIN191BLUE SLIN194IWBLUE SLIN192BLUE
Sterilin® Dippas® Milk Dippers
Probenehmer Probenschöpfer
These sample dippers facilitate the sample collection and subsequent transportation to the laboratory in a single container with no risk of cross contamination.
  • Individually wrapped and gamma irradiated to ensure sterility
  • Available in either clear or blue PS for the food industry
  • Handle neatly snaps off after sample collection for ease of transportation
Zertifizierungen: EN 14245 and BS5213.
Lieferumfang: Supplied with a 'flow-seal' screw cap for a leak tight seal.
Bitte beachten *Produkte auf dieser Seite sind außerhalb des AnwendungsbereichsMore Product Information