Digital thermo-hygrometer
Lieferant: Industrial Physics
RV1610
SIMERV1610EA 55.7 CHF
SIMERV1610
Digital thermo-hygrometer
Hygrometer Thermohygrometer, digital
This handheld thermo-hygrometer is designed to measure temperature and humidity with readable surronding distance of ten meters.
  • Large display (56×40 mm)
  • Toggle between °C/°F
  • Maximum/minimum values
  • Time display and alarm
  • Memory function