HyClone™ Earle's 1X Balanced Salt Solution (EBSS): Solution
Lieferant: HyClone products (Cytiva)
SH30029.02
SH30029.02EA 33.8 CHF
SH30029.02
HyClone™ Earle's 1X Balanced Salt Solution (EBSS): Solution
Puffer Gewebekultur-Puffer
Balanced salt solution for cell culture applications.
  • All formulations incorporate a rigorous specification and approval process
  • Available in lot sizes that facilitate scale-up and manufacturing
  • Endotoxin Concentration 1.0 EU/mL
Zertifizierungen: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Bitte beachten Shelf life 24 months. Storage 15° to 3° C