SGE OptChem™ ConnecTite Inlet Liners
Lieferant: Trajan Scientific and Medical
0926232401 09262324 0926232501 09262325 0926232601 09262327
SGEA0926232401EA 220 CHF
SGEA0926232401 SGEA09262324 SGEA0926232501 SGEA09262325 SGEA0926232601 SGEA09262327
SGE OptChem™ ConnecTite Inlet Liners
Chromatographie-Zubehör Chromatographie-Liner
The SGE inlet liner range aims to make it simple for all gas chromatographers to select the correct liner.
  • Easy to choose - colour coded by geometry to simplify your selection
  • Easy to use - contains O-rings so ready to go
  • Confidence in your analysis - certified deactivation
  • Robust and durable for longer life, less maintenance downtime and reduced maintenance costs
  • Color coded orange
  • Analytical formatted
ConnecTite liners facilitate maximum transfer of sample to the GC column and inhibit sample degradation by reducing contact with hot metal componentsMore Product Information