Jars, wide mouth, with screw cap
Lieferant: SEMADENI
3742
SEMA3742EA 6.6 CHF
SEMA3742
Jars, wide mouth, with screw cap
Dosen Dosen, Universal
Jars, wide mouth, made of HDPE, white.
  • Space saving
  • With inner seal and red screw cap
  • Good chemical and temperature resistance