Sterile-venting membrane screw caps, DURAN®
Lieferant: DWK Life Sciences
291182809 291181307 291181907 291185609 291189105 291162807
SCOT291182809EA 50.1 CHF
SCOT291182809 SCOT291181307 SCOT291181907 215-4260 SCOT291189105 SCOT291162807
Sterile-venting membrane screw caps, DURAN®
Verschlüsse Flaschenverschlüsse
Ideal for autoclaving processes because the 0,2 micron ePTFE membrane permits pressure equalisation and tight sealing, greatly reducing the risk of contamination. Ingress of liquids or solids is prevented and the bottle contents remain sterile.
  • Membrane caps can be autoclaved at 121 °C (20 minutes) or 134 °C (3 minutes)
  • Cost effective as the caps can be reused several times
  • Convenient as six caps are available allowing the use with a large range of bottle sizes from 10 ml up to 3500 ml
  • Biocompatible: The ePTFE filter material conforms to USP 88 121 °C
More Product Information