Redox combination electrodes, AquaLine
Lieferant: SI Analytics
285113073
SCOI285113073EA 323 CHF
SCOI285113073
Redox combination electrodes, AquaLine
Elektroden Redox-Elektroden
AquaLine electrodes are ideal for measuring applications in drinking water, industrial water and sewage range.