Pocket thermometers
Lieferant: Ludwig Schneider GmbH & CO. KG
1481265 1481365 1481562 1481572 1481573 1481574 1021142
SCNL1481265EA 17.7 CHF
SCNL1481265 SCNL1481365 SCNL1481562 SCNL1481572 SCNL1481573 SCNL1481574 SCNL1021142
Pocket thermometers
Thermometer Analoge Thermometer
These pocket thermometers are protected designs (DBGM). Mercury, red or blue filling.
  • Total immersion
  • Available in different capillary forms
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.