EPA vials with PP cap
Lieferant: DWK Life Sciences
SP60961C-4
SCERSP60961C-4EA 556 CHF
SCERSP60961C-4
EPA vials with PP cap
Vials
These clear screw thread EPA vials are used for the collection and storage of water samples.
  • Not pre-cleaned
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Zertifizierungen: These vials comply with the guidelines for establishing test procedures for the analysis of pollutants (Ref. EPA Methods 601, 602, 603 and 604).
Verpackung: Vials are packaged in a plastic shrink wrap module, lehr-clean, as they come from the end of the production lehr and are placedMore Product Information
Order Now
TECHNISCHE DATEN
Environmentally Preferable