Stopper, HDPE, KIMBLE®
Lieferant: DWK Life Sciences
28160R-19 SP28160R-13
KIML28160R-19EA 0 CHF
KIML28160R-19 SCERSP28160R-13
Stopper, HDPE, KIMBLE®
Stopfen PE/LDPE/HDPE-Stopfen
HDPE, linear, ideal for flasks that are grounded to [ST] stopper dimensions.
  • Yellow and blue stopper with a closed bottom
  • Enlarged flange is designed to protect the neck if the glassware is tipped over