ThermoSafe® Nomadic® Pre-Qualified 2 to 8 °C Shippers
Lieferant: Sonoco Thermosafe
E280C
SCATE280CEA 461 CHF
SCATE280C
ThermoSafe® Nomadic® Pre-Qualified 2 to 8 °C Shippers
Versandtaschen und Transportbehälter
Nomadic® pre-qualified shippers are designed for the shipment of pharmacetuicals, biological specimens, and samples.
  • Pre-qualified 2 to 8°C shipper which delivers 48 hour performance
  • PUR insulation base and lid in robust outer corrugate
  • Qualified duration: 48 hours
  • Ideal for use as diagnostic specimen and infectious substance compliant shipper overpacks
Nomadic® was the industry's first pre-qualified solution and is still trusted for pharmaceutical and clinical trial products around the world. Nomadic® shippers can be used to ship minimum loads of 1More Product Information
Order Now
TECHNISCHE DATEN
Environmentally Preferable