ThermoSafe® PUR Insulated Shippers
Lieferant: Sonoco Thermosafe
E126 E38HTOP E337 E186 E327 E366 E145
SCATE126EA 157 CHF
SCATE126 SCATE38HTOP SCATE337 SCATE186 SCATE327 SCATE366 SCATE145
ThermoSafe® PUR Insulated Shippers
Versandtaschen und Transportbehälter
High perfomance polyurethane (PUR) insulated shippers with foam plug closure.
  • PUR offers greater insulating capability when compared to same size EPS container
  • Thick walls and outer corrugate provides protective shock absorption and cushioning during shipping and handling
  • Protects highly temperature-sensitive shipments
  • Economical and lightweight
Rigid, one-piece, three-layer laminate construction provides excellent durability. Proprietary process laminates insulation core to inner and outer corrugate walls for superior strength and temperature controlMore Product Information