Cleaning cloth, low-lint, Tork Industrial
Lieferant: Essity
190578
SCAA190578EA 453 CHF
SCAA190578
Cleaning cloth, low-lint, Tork Industrial
Wischtücher Wisch- und Handtücher, Einweg Wischtücher für sensible Anwendungen
This extremely low lint cloth with anti-static properties is ideal for use with solvents when cleaning smooth surfaces. Made from 100% PP.
  • Perfect for working with sensitive surfaces due to extreme the low-lint and antistatic properties
  • Strong texture for excellent oil and water absorption
  • Tork Easy Handling™ for easier carrying, opening and disposing of packaging
  • The silicone-free cloth ensures that there are no residues on a surface
  • Food contact approved
Suitable for use in clean condition areas, printing industry, precision engineering, electronicsMore Product Information