X-ray and metal detectable micro spatulas
Lieferant: Sampling Systems
M1013B-170
SAMPM1013B-170EA 15.1 CHF
SAMPM1013B-170
X-ray and metal detectable micro spatulas
Spatel
The X-ray and metal detectable micro spatulas are ideal for use in food and pharmaceutical production and quality control work. The spatulas are made from a special grade of PP that can be detected. The bright blue colour also means that they can be easily seen if left in the powder in line with HACCP guidelines.
  • Robust PP construction
  • PP gives excellent chemical resistance to many products
  • Long ergonomic handle
  • Full batch traceability
The spatula are ideal for sampling from bags and surface sampling from containers aswell as general work. The narrowMore Product Information