Donkey serum
Lieferant: Biowest
S2170-100 S2170-500
S2170-100EA 79.8 CHF
S2170-100 S2170-500
Donkey serum
Seren
Donkey serum may be used as a blocking agent and controls in immunoassays. For immunohistochemistry and immunocytochemistry, donkey serum was used between 5 to 20% as a blocking agent in various studies.