Respirators with FR chin seal, Z-Link Series
Lieferant: RPB SAFETY
16-010-31-CE 16-670 16-810 16-811 16-531 16-522
RPBS16-010-31-CEEA 0 CHF
RPBS16-010-31-CE RPBS16-670 RPBS16-810 RPBS16-811 RPBS16-531 RPBS16-522
Respirators with FR chin seal, Z-Link Series
Atemschutzmasken Atemschutz-Systeme
The RPB Z-link respirator with FR chin seal, is the world's most comfortable (fully padded) and versatile industry respirator. Providing the highest respiratory protection available (TH3), the Z-Link system also provides hard hat protection, eye and face protection and hearing protection.
  • Visor hinges open for easy communication with team (outside of respiratory hazards)
  • Air flow and distribution is fully adjustable with the air deflector system to suit personal preferences
  • Fixed jaw ensures secure cape attachment
  • Vision-Link™ light mount point
  • ANSI Z89.1