Kaliumacetat

Lieferant: Roth Carl

Synonyms: Acetic acid potassium salt

T874.1 T874.2 T874.5
ROTHT874.1EA 51 CHF
ROTHT874.1 ROTHT874.2 ROTHT874.5
Kaliumacetat
Kaliumacetat
Formel: CH₃COOK
MW: 98.14 g/mol
Melting Pt: 304
Dichte: 1.57 g/cm³ (25 °C)
Flash-Pt: >250 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00012458
CAS-Nummer: 127-08-2
EINECS: 204-822-2
Merck Index: 13,07687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR