Natriumacetat Trihydrat

Lieferant: Roth Carl

Synonyms: Acetic acid sodium salt trihydrate

6779.1 6779.2 6779.3 6779.5
ROTH6779.1EA 77.9 CHF
ROTH6779.1 ROTH6779.2 ROTH6779.3 ROTH6779.5
Natriumacetat Trihydrat
Natriumacetat Trihydrat
Formel: H₃CCOONa·3H₂O
MW: 136.08 g/mol
Siedepunkt: 120…123 °C (1013 hPa)
Melting Pt: 58
Dichte: 1.4 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00071557
CAS-Nummer: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR