TRITON™ X-100 (Polyethylenglycol-tert-octylphenylether)

Lieferant: Roth Carl
Gefahr

Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol

6683.1 6683.2
ROTH6683.1EA 0 CHF
ROTH6683.1 ROTH6683.2
TRITON™ X-100 (Polyethylenglycol-tert-octylphenylether)
TRITON™ X-100 (Polyethylenglycol-tert-octylphenylether)
Siedepunkt: ∼200 °C (1013 hPa)
Melting Pt: 6 °C
Dichte: 1.06 g/cm³ (20 °C)
Flash-Pt: 251 °C (closed cup)
Lager Temperatur: Kühlschrank
MDL Number: MFCD00132505
CAS-Nummer: 9002-93-1
UN: 3082
ADR: 9,III
Merck Index: 13,06793

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR