Magnesiumacetat Tetrahydrat

Lieferant: Roth Carl

Synonyms: Acetic acid magnesium salt tetrahydrate, Magnesium di(acetate) tetrahydrate

0275.1 0275.2 0275.3
ROTH0275.1EA 42.6 CHF
ROTH0275.1 ROTH0275.2 ROTH0275.3
Magnesiumacetat Tetrahydrat
Magnesiumacetat Tetrahydrat
Formel: (H₃CCOO)₂Mg·4H₂O
Melting Pt: 80 °C
Dichte: 1.454 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149214
CAS-Nummer: 16674-78-5
EINECS: 205-554-9
Merck Index: 13,05676

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR