Competent cell buffer
Lieferant: Rockland Immunochemicals
MB-057
ROCKMB-057EA 515 CHF
ROCKMB-057
Competent cell buffer
Puffer Klonpuffer
This product is ready-to-use as a working 1X solution and requires no further dilution. This buffer consists of 60 mM Calcium Chloride, 15% v/v Glycerol, 10mM Tris HCl at a pH of 7,5.
Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade deionised water.
This product was aseptically filtered through a Millipore 0,22 µm filter into clean, pre-sterilised containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Bitte beachten This product is for research use only and is not intended for therapeutic or diagnostic applications.