TrueBlot® IP/Western Blot

Lieferant: Rockland Immunochemicals
13-8817-82 00-8811-25 13-8817-80 88-1488-31 88-1688-31 88-7788-31
ROCK13-8817-82EA 488 CHF
ROCK13-8817-82 ROCK00-8811-25 ROCK13-8817-80 ROCK88-1488-31 ROCK88-1688-31 ROCK88-7788-31
TrueBlot® IP/Western Blot
Blotting-Kits
IP Western blots provide highly specific results, yet often suffer from heavy/light chain blotting, contamination, and ongoing interference.

TrueBlot® reagents enable you to generate clear, best-quality data in your Immunoprecipitation and Western Blot protocols. Available in several options, from IP Beads alone, to complete IP/Western Blot kits from goat, mouse, rabbit or sheep.

TrueBlot® products solve nearly all of these problems through increased sensitivity, less background noise, and enhanced accuracy.
Order Now

TECHNISCHE DATEN

Environmentally Preferable
Konzentration 14,4 mg/cc antibody per cc agarose 0,5 mg/ml by UV absorbance at 280 nm
Kreuzadsorption No
Species reactivity Mouse Mouse Goat Rabbit Mouse
Transporttemperatur Wet Ice Wet Ice Wet Ice Wet Ice Wet Ice
Lagerungspuffer 0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, Proprietary 0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, Proprietary 0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, Proprietary 0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, Proprietary 0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, Proprietary
Lagertemperatur 4 °C 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR