Anti-IgG F(c) Goat Polyclonal Antibody (Peroxidase)

Lieferant: Rockland Immunochemicals
712-1322
ROCK712-1322EA 507 CHF
ROCK712-1322
Anti-IgG F(c) Goat Polyclonal Antibody (Peroxidase)
Antikörper
This product has been assayed against 1.0 µg of Rat IgG in a standard capture ELISA using ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid)) as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:10000 - 1:100000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 500 µl
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

TECHNISCHE DATEN

Antigen-Symbol IgG F(c)
Environmentally Preferable
Antikörper-Typ Sekundär
Klonalität Polyklonal
Konjugation Peroxidase
Reaktivität Rat
Wirt Goat
Isotyp
Western blot Yes
ELISA Yes
ImmunoChemie Yes
Immunpräzipitation Yes
Gelverschiebung Yes
Durchflusszytometrie Yes
ChIP Yes
Neutralisation Yes
Antikörperfragment F(ab')2
Immunogen Rat IgG F(c) fragment
Format Antibody fraction
Form Lyophilized
Reinigung purified by immunoaffinity chromatography
Konzentration 1,0 mg/ml by UV absorbance at 280 nm
Kreuzadsorption Bovine, Horse, Human
Lagerungspuffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Gentamicin Sulfate. 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Lagertemperatur Cool
Transporttemperatur Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR