Anti-IgA Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Rockland Immunochemicals
618-105-006
ROCK618-105-006EA 544 CHF
ROCK618-105-006
Anti-IgA Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Secondary Goat Anti-IgA Reacts with Ferret

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:3000; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgA α
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgA
Reactivity: Ferret
Order Now

TECHNISCHE DATEN

Antigen-Symbol IgA
Environmentally Preferable
Antikörper-Typ Sekundär
Klonalität Polyklonal
Konjugation AP (Alkaline Phosphatase)
Reaktivität Ferret
Wirt Goat
Isotyp
Western blot Yes
ELISA Yes
ImmunoChemie Yes
Immunpräzipitation Yes
Gelverschiebung Yes
Durchflusszytometrie Yes
ChIP Yes
Neutralisation Yes
Immunogen Ferret IgA alpha heavy chain
Format Antibody fraction
Form Liquid (sterile filtered)
Reinigung purified by immunoaffinity chromatography
Konzentration 1,0 mg/ml by UV absorbance at 280 nm
Kreuzadsorption none
Lagerungspuffer 0,05 M Tris Chloride, 0,15M Sodium Chloride, 0,001M Magnesium Chloride, 0,0001M Zinc Chloride, 50% (v/v) Glycerol
Lagertemperatur Cool
Transporttemperatur Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR