Anti-IgG F(ab')2 Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Rockland Immunochemicals
610-1504
ROCK610-1504EA 465 CHF
ROCK610-1504
Anti-IgG F(ab')2 Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
This product has been assayed against 1.0 µg of Mouse IgG in a standard capture ELISA using pNPP p-nitrophenyl phosphate code # NPP-10 as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

TECHNISCHE DATEN

Antigen-Symbol IgG F(ab')2
Environmentally Preferable
Antikörper-Typ Sekundär
Klonalität Polyklonal
Konjugation AP (Alkaline Phosphatase)
Reaktivität Mouse
Wirt Goat
Isotyp
Western blot Yes
ELISA Yes
ImmunoChemie Yes
Immunpräzipitation Yes
Gelverschiebung Yes
Durchflusszytometrie Yes
ChIP Yes
Neutralisation Yes
Immunogen Mouse IgG F(ab')2 fragment
Format Antibody fraction
Form Liquid (sterile filtered)
Reinigung purified by immunoaffinity chromatography
Konzentration 0,76 mg/ml by UV absorbance at 280 nm
Kreuzadsorption none
Lagerungspuffer 0,05 M Tris Chloride, 0,15M Sodium Chloride, 0,001M Magnesium Chloride, 0,0001M Zinc Chloride, 50% (v/v) Glycerol
Lagertemperatur Cool
Transporttemperatur Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR