Anti-IgA Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
609-4106
ROCK609-4106EA 337 CHF
ROCK609-4106
Anti-IgA Rabbit Polyclonal Antibody
Antikörper
Secondary Rabbit Anti-IgA (alpha chain) Reacts with Human

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:250 - 1:1000;

Type: Secondary
Antigen: IgA α
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgA
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR