Anti-KLH Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))

Lieferant: Rockland Immunochemicals
600-402-466
ROCK600-402-466EA 899 CHF
ROCK600-402-466
Anti-KLH Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Antikörper
Recommended Dilutions:
Restore with deionized water (or equivalent); 100 µl

Type: Tag
Antigen: KLH
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR