Anti-ORAI1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
600-401-C00
ROCK600-401-C00EA 1100 CHF
ROCK600-401-C00
Anti-ORAI1 Rabbit Polyclonal Antibody
Antikörper
ORAI1 antibody has been tested in ELISA, WB, and IHC. ORAI1 often migrates at a higher than expected (~32 kDa) molecular weight in SDS-PAGE. Expect a band at ~51 kDa in size corresponding to ORAI1 by western blotting in the appropriate cell lysate.

Recommended Dilutions: ELISA: 1:10000 - 1:40000; Western Blot: 0.5 - 1.0 µg/ml; Immunochemistry: 10 µg/ml;

Type: Primary
Antigen: ORAI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR