Anti-GFP Mouse Polyclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 9F9.F9]

Lieferant: Rockland Immunochemicals
600-303-215
ROCK600-303-215EA 1700 CHF
ROCK600-303-215
Anti-GFP Mouse Polyclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 9F9.F9]
Antikörper
Monoclonal anti-GFP is designed to detect enhanced GFP and GFP containing recombinant Proteins. This antibody can be used to detect GFP by ELISA (sandwich or capture) for the direct binding of antigen.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:2000 - 1:5000; Immunochemistry: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Tag
Antigen: GFP
Clonality: Polyclonal
Clone: 9F9.F9
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Full length
Host: Mouse
Isotype: IgG1 kappa
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR