Anti-IgG (H+L) Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
308-1102
ROCK308-1102EA 570 CHF
ROCK308-1102
Anti-IgG (H+L) Goat Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Horse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR