Anti-CCL20 Mouse Monoclonal Antibody (Unconjugated) [clone: 3N2D9]

Lieferant: Rockland Immunochemicals
209-301-B51
ROCK209-301-B51EA 977 CHF
ROCK209-301-B51
Anti-CCL20 Mouse Monoclonal Antibody (Unconjugated) [clone: 3N2D9]
Antikörper
MIP-3a antibody has been tested for use in ELISA, IP, and Neutralization. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 0.5µg/ml;

Type: Primary
Antigen: CCL20
Clonality: Monoclonal
Clone: 3N2D9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR