Anti-6xHis Mouse Monoclonal Antibody (Unconjugated) [clone: 33D10.D2]

Lieferant: Rockland Immunochemicals
200-301-382S
ROCK200-301-382SEA 263 CHF
ROCK200-301-382S
Anti-6xHis Mouse Monoclonal Antibody (Unconjugated) [clone: 33D10.D2]
Antikörper
Anti-6X His is optimally suited for monitoring expression of His-tagged fusion Proteins. As such, anti-6X His/6X His can be used to identify fusion Proteins that contain the 6X His epitope.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunoprecipitation: 1:200;

Type: Tag
Antigen: 6xHis
Clonality: Monoclonal
Clone: 33D10.D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR