Western blot kits, TrueBlot®

Lieferant: Rockland Immunochemicals

18-8814-31 18-8815-31 18-8814-33 18-8815-33 18-8816-31 18-8816-33 18-8817-30 18-8817-31 18-8817-33 88-8884-31 88-8886-31 88-8887-31
ROCK18-8814-31EA 280 CHF
ROCK18-8814-31 ROCK18-8815-31 ROCK18-8814-33 ROCK18-8815-33 ROCK18-8816-31 ROCK18-8816-33 ROCK18-8817-30 ROCK18-8817-31 ROCK18-8817-33 ROCK88-8884-31 ROCK88-8886-31 ROCK88-8887-31
Western blot kits, TrueBlot®
Elektrophorese-Reagenzien Western Blotting Reagents

TrueBlot® Western blot kits contain the critical supporting reagents, buffers, and substrates for immunoprecipitation and Western blotting of samples using TrueBlot® second step immunoblotting reagents in conjunction with the user's own primary IP antibody and primary (mouse, goat or rabbit IgG) Western blotting antibody.


TrueBlot® technology enables unhindered detection of protein bands of interest which would otherwise be obscured by the presence of reduced and denatured heavy and light chain immunoglobulin in the blot (as detected by the conventional immunoblotting HRP IgG reagents). TrueBlot® is the unique horseradish peroxidase conjugated anti-mouse/anti-goat/anti-rabbit IgG immunoblotting second step reagent which enables detection of immunoblotted target protein bands, without hindrance by interfering immunoglobulin heavy and light chains from the user's own IP antibody. TrueBlot® preferentially detects the non reduced form of IgG over the reduced, SDS-denatured form of IgG.


Applications include studies examining post-translational modification (e.g. phosphorylation or acetylation) or protein-protein interactions.


Lieferumfang: Contains sufficient reagents for up to 20 blots.


Bitte beachten For research use only and not recommended for therapeutic or diagnostic applications.

Order Now

TECHNISCHE DATEN

Environmentally Preferable
Konzentration 1,0 mg/ml by UV absorbance at 280 nm
Transporttemperatur Wet ice
Lagertemperatur −20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR