Anti-TNFA Rabbit Monoclonal Antibody (Unconjugated) [clone: IPO-38]

Lieferant: Rockland Immunochemicals
110-401-321
ROCK110-401-321EA 2390 CHF
ROCK110-401-321
Anti-TNFA Rabbit Monoclonal Antibody (Unconjugated) [clone: IPO-38]
Antikörper
Primary Rabbit Anti-Mouse TNF alpha Reacts with Mouse

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000;

Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone: IPO-38
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR