Anti-PRSS2 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
100-4180
ROCK100-4180EA 2390 CHF
ROCK100-4180
Anti-PRSS2 Rabbit Polyclonal Antibody
Antikörper
Primary Rabbit Anti-TRYPSINOGEN (Bovine Pancreas) Reacts with Pan

Recommended Dilutions: ELISA: 1:10000 - 1:40000; Western Blot: 1:1000 - 1:4000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: PRSS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR