Anti-CA2 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
100-4157
ROCK100-4157EA 2390 CHF
ROCK100-4157
Anti-CA2 Rabbit Polyclonal Antibody
Antikörper
This product has been assayed against 1.0 µg of Carbonic Anhydrase II (Bovine Erythrocytes) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat) and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: CA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR