Anti-NFYA Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
100-401-100
ROCK100-401-100EA 954 CHF
ROCK100-401-100
Anti-NFYA Rabbit Polyclonal Antibody
Antikörper
Anti-NF-Y (A subunit) Antibody was assayed by immunoblot and found to be reactive against NF-Y (A subunit specific) at a dilution of 1:6,000 followed by reaction with Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat).

Recommended Dilutions: ELISA: 1:20000 - 1:100000; GelShift: Use 2.0 µl; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:400;

Type: Primary
Antigen: NFYA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR