Environmental Monitoring Solutions, RmoniWeb Validated
Lieferant: Rmoni
BUZZ.LIGHT PT100L150SIL DSWL070ALU NTCL090PVC RMZ1215R34 RMZ1215G13 RMZ1215E13 RMZ1215E23 RMZ1215E30 RMZ1215E33 RMZ1215R40 RMZ1215R43 RMZ1215R51 RHL050EE2.5AN DPSI250L025 RMZ1214E51
620-2842EA 71.6 CHF
620-2842 620-2846 620-2844 620-2845 620-2856 620-2855 620-2849 620-2850 620-2851 620-2852 620-2857 620-2858 620-2859 620-2848 620-2843 RMONRMZ1214E51
Environmental Monitoring Solutions, RmoniWeb Validated
Überwachungssysteme CO2- und Temperatur-Überwachungssysteme
Rmoni offers complete end-2-end sensor solutions to set up an Environmental Monitoring Solutions (EMS) in hospitals, laboratories, pharmacies and cleanrooms. Keeping track of all critical parameters including temperature, humidity and many more. Ensuring product and process integrity during storage, handling and as well as transportation.
  • Integrated EMS solutions for labs, healthcare and pharma
  • Sensors, digital checklists and professional services (including support and service level agreements, (re-)calibration, installation and training)
  • FDA 21 part 11
More Product Information
Order Now
TECHNISCHE DATEN
Environmentally Preferable