Iodmonobromid 20 g/l in Essigsäure 99% Reag. Ph. Eur. 1043504

Lieferant: REAGECON
Gefahr

1045901
REGE1045901EA 417 CHF
REGE1045901
Iodmonobromid 20 g/l in Essigsäure 99% Reag. Ph. Eur. 1043504
Iodmonobromid
Formel: IBr
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011353
CAS-Nummer: 7789-33-5
EINECS: 232-159-9
UN: 2789
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR